missing translation for 'onlineSavingsMsg'
Learn More

Triisopropyl orthoformate, 95%

Product Code. 10441691 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10441691 25 mL Glass bottle 25mL
10646184 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10441691 Supplier Thermo Scientific Acros Supplier No. 325440250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 4447-60-3 | C10H22O3 | 190.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4447-60-3
Molecular Formula C10H22O3
Molecular Weight (g/mol) 190.29
MDL Number MFCD00060628
InChI Key FPIVAWNGRDHRSQ-UHFFFAOYSA-N
Synonym triisopropyl orthoformate, triisopropoxymethane, triisopropylorthoformate, propane, 2,2',2-methylidynetris oxy tris, 2-diisopropoxymethoxy propane, 2-di propan-2-yloxy methoxy propane, 2-bis propan-2-yloxy methoxy propane, orthoformic acid, triisopropyl ester, isopropyl orthoformate
PubChem CID 78191
IUPAC Name 2-[di(propan-2-yloxy)methoxy]propane
SMILES CC(C)OC(OC(C)C)OC(C)C

Specifications

Color Colorless
Density 0.8600g/mL
Boiling Point 64°C to 65°C (18.0 mmHg)
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula HC(OCH(CH3)2)3
Refractive Index 1.3950 to 1.398
Quantity 25 mL
Specific Gravity 0.86
Formula Weight 190.29
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Triisopropyl orthoformate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.