missing translation for 'onlineSavingsMsg'
Learn More

Triisopropyl Borate, TRC

Product Code. 30452008
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452008 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30452008 Supplier Toronto Research Chemicals Supplier No. TRCT795490100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triisopropyl Borate
CAS 5419-55-6
Molecular Formula C9H21BO3
Quantity 100 g
Synonym Boric Acid (H3BO3) Triisopropyl Ester, Isopropyl Borate, Boric Acid Triisopropyl Ester, Boron Isopropoxide, Boron Triisopropoxide, Isopropyl Borate ((C3H7O)3B), NSC 9779, Triisopropoxyborane, Triisopropoxyboron, Triisopropyl Borate, Triisopropyl Orthoborate, Tris(1-methylethyl) Borate, Trisisopropoxyborane
InChI Formula InChI=1S/C9H21BO3/c1-7(2)11-10(12-8(3)4)13-9(5)6/h7-9H,1-6H3
SMILES CC(C)OB(OC(C)C)OC(C)C
IUPAC Name tripropan-2-yl borate
Molecular Weight (g/mol) 188.07
Formula Weight 188.1584
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.