missing translation for 'onlineSavingsMsg'
Learn More

Triisopropanolamine, 98%, Thermo Scientific Chemicals

Product Code. 10479930
Click to view available options
Quantity:
5 g
100 g
1 kg
10 kg
Packaging:
Glass bottle
Plastic Can
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-20-3
Molecular Formula C9H21NO3
Molecular Weight (g/mol) 191.27
MDL Number MFCD00004533
InChI Key SLINHMUFWFWBMU-UHFFFAOYSA-N
Synonym triisopropanolamine, tipa, tri-2-propanolamine, tri-iso-propanolamine, tris 2-propanol amine, 2-propanol, 1,1',1-nitrilotris, tris 2-hydroxypropyl amine, 1,1',1-nitrilotripropan-2-ol, tris 2-hydroxy-1-propyl amine, caswell no. 891
PubChem CID 24730
IUPAC Name 1-[bis(2-hydroxypropyl)amino]propan-2-ol
SMILES CC(CN(CC(C)O)CC(C)O)O

Specifications

Melting Point 48.0°C to 52.0°C
Color White to Yellow
Boiling Point 190.0°C (23.0 mmHg)
Flash Point 160°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula N(CH2CH(OH)CH3)3
Quantity 5 g
Beilstein 04,762
Fieser 17,371; 12,445; 13,46
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol, diethyl ether
Viscosity 138 mPa.s (60°C)
Formula Weight 191.27
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Triisopropanolamine
Show More Show Less
compliance-icons
Product Identifier
  • Triisopropanolamine
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'