missing translation for 'onlineSavingsMsg'
Learn More

Triisobutylaluminium, 1.0M solution in toluene, AcroSeal™

Product Code. 10301753 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10301753 800 mL AcroSeal™ Glass Bottle 800mL
10178692 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10301753 Supplier Thermo Scientific Acros Supplier No. 185428000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 100-99-2 | C12H27Al | 198.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-99-2 , 108-88-3
Molecular Formula C12H27Al
Molecular Weight (g/mol) 198.33
MDL Number MFCD00008929
InChI Key MCULRUJILOGHCJ-UHFFFAOYSA-N
Synonym triisobutylaluminum, triisobutylaluminium, aluminum, tris 2-methylpropyl, tibal, triisobutylalane, triisobutyl aluminum, aluminum, triisobutyl, tris isobutyl alane, tris isobutyl aluminum, unii-09p2thv2x4
PubChem CID 16682931
IUPAC Name tris(2-methylpropyl)alumane
SMILES CC(C)C[Al](CC(C)C)CC(C)C

Specifications

Color Colorless to Yellow
Linear Formula [(CH3)2CHCH2]3Al
Fieser 09,486
Density 0.8400g/mL
Merck Index 10, 316
Solubility Information Solubility in water: reacts. Other solubilities: miscible with aromatic and aliphatic hydrocarbons
Formula Weight 198.33
Percent Purity 24.5 to 25.5% (on Al basis)
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.84
Quantity 800 mL
Flash Point 4°C
Physical Form Solution
Chemical Name or Material Triisobutylaluminium, 1.1M Solution in Toluene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Triisobutylaluminium, solution in toluene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.