missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Triglycerides Reagent

Product Code. 17049193 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
2 x 125 mL
Unit Size:
320g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17049193 2 x 125 mL 320g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17049193 Supplier Thermo Scientific™ Supplier No. TR22421

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Determine quantity of triglycerides in human serum or plasma in vitro with Thermo Scientific™ Triglycerides Reagent.

Measurement of triglycerides is important in the diagnosis and management of hyperlipidaemias. Levels may be increased in nephrotic syndrome, coronary artery disease (CAD), hypothyroidism, diabetes mellitus and liver disease. Decreased values are sometimes observed in cases of protein malnutrition, hyperthyroidism and cachectic states.
TRUSTED_SUSTAINABILITY

Specifications

Description Infinity Triglyceride 2 x 125mL
Detectable Analytes Triglycerides
Sample Type Plasma, Serum
Type Reagent
Linearity Up to 10mmol/L (885mg/dL)
Methods Compatibility Endpoint
Quantity 2 x 125 mL
Stability 18 months at 2-8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.