missing translation for 'onlineSavingsMsg'
Learn More

Triflusal, MedChemExpress

Product Code. 16448518
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448518 10 mg 10mg
16434387 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448518 Supplier Medchem Express Supplier No. HYB0531
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Triflusal irreversibly inhibits the production of thromboxane-B2 in platelets by acetylating cycloxygenase-1.

Triflusal irreversibly inhibits the production of thromboxane-B2 in platelets by acetylating cycloxygenase-1.

Specifications

Chemical Name or Material Triflusal
CAS 322-79-2
Color White
Purity Grade Notes Research
Molecular Formula C10H7F3O4
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (402.97 mM; Need ultrasonic)
SMILES O=C(O)C1=CC=C(C(F)(F)F)C=C1OC(C)=O
Molecular Weight (g/mol) 248.16
Formula Weight 248.16
Percent Purity 99.64%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.