Learn More
MedChemExpress Trifluridine/tipiracil hydrochloride mixture
Description
The composition of Trifluridine/tipiracil hydrochloride mixture (TAS-102) is a 1:0.5 mixture (on a molar basis) of alpha,alpha,alpha-tri-fluorothymidine (FTD) and thymidine phosphorylase inhibitor (TPI). Trifluridine/tipiracil hydrochloride mixture (TAS-102) shows the antitumor activity mainly via the inhibition of thymidylate synthase (TS) and incorporation into DNA.
Specifications
Specifications
| Molecular Weight (g/mol) | 435.76 g/mol |
| Product Type | Reference compound |
| Content And Storage | 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture) |
| Shipping Condition | Room temperature |
| Purity | 98.0% |
| Quantity | 10 mg |
| Solubility Information | H2O : 100 mg/mL (229.48 mM∣Need ultrasonic) ∣DMF : 20 mg/mL (45.90 mM∣Need ultrasonic) ∣DMSO : 2.34 mg/mL (5.37 mM∣Need ultrasonic and warming) |
| For Use With (Application) | Cancer-programmed cell death |
| CAS | 733030-01-8 |
| Synonym | TAS-102 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.