missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Trifluridine/tipiracil hydrochloride mixture

Product Code. 16423107
Encompass_Preferred
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 16423107

Brand: MedChemExpress HY16478

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Trifluridine/tipiracil hydrochloride mixture (TAS-102) is a potent and orally active nucleoside antitumor agent.

The composition of Trifluridine/tipiracil hydrochloride mixture (TAS-102) is a 1:0.5 mixture (on a molar basis) of alpha,alpha,alpha-tri-fluorothymidine (FTD) and thymidine phosphorylase inhibitor (TPI). Trifluridine/tipiracil hydrochloride mixture (TAS-102) shows the antitumor activity mainly via the inhibition of thymidylate synthase (TS) and incorporation into DNA.

Specifications

Molecular Weight (g/mol) 435.76 g/mol
Product Type Reference compound
Content And Storage 4°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Shipping Condition Room temperature
Purity 98.0%
Quantity 50 mg
Solubility Information H2O : 100 mg/mL (229.48 mM∣Need ultrasonic) ∣DMF : 20 mg/mL (45.90 mM∣Need ultrasonic) ∣DMSO : 2.34 mg/mL (5.37 mM∣Need ultrasonic and warming)
For Use With (Application) Cancer-programmed cell death
CAS 733030-01-8
Synonym TAS-102
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.