missing translation for 'onlineSavingsMsg'
Learn More

Trifluridine, MedChemExpress

Product Code. 16421356
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16421356 200 mg 200mg
16418016 100 mg 100mg
16404177 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16421356 Supplier Medchem Express Supplier No. HYA0061
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Trifluridine (Trifluorothymidine; 5-Trifluorothymidine; TFT) is an irreversible thymidylate synthase inhibitor, and thereby suppresses DNA synthesis. Trifluridine is an antiviral drug for herpes simplex virus (HSV) infection. Trifluorothymidine also has anti-orthopoxvirus activity.

Trifluridine (Trifluorothymidine

Specifications

Chemical Name or Material Trifluridine
Physical Form Solid
CAS 70-00-8
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C10H11F3N2O5
For Use With (Application) Cancer-programmed cell death
Synonym Trifluorothymidine 5-Trifluorothymidine TFT
Solubility Information DMSO : ≥ 100 mg/mL (337.61 mM)
SMILES O=C1C(C(F)(F)F)=CN([C@@H]2O[C@H](CO)[C@@H](O)C2)C(N1)=O
Molecular Weight (g/mol) 296.2
Formula Weight 296.2
Percent Purity 99.14%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.