missing translation for 'onlineSavingsMsg'
Learn More

Triflupromazine Hydrochloride, Mikromol™

Product Code. 18001990
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18001990 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18001990 Supplier Mikromol Supplier No. MM1675.00
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Triflupromazine Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Triflupromazine hydrochloride
CAS 1098-60-8
Analyte or Component Names Triflupromazine hydrochloride
Shipping Condition Room Temperature
Molecular Formula C18 H19 F3 N2 S . Cl H
Certifications/Compliance ISO 17025
Synonym Triflupromazine Hydrochloride, 10H-Phenothiazine-10-propanamine, N,N-dimethyl-2-(trifluoromethyl)-, monohydrochloride (9CI), Phenothiazine, 10-(3-dimethylaminopropyl)-2-(trifluoromethyl)-, hydrochloride (6CI), Phenothiazine, 10-[3-(dimethylamino)propyl]-2-(trifluoromethyl)-, monohydrochloride (8CI), 10-(3-Dimethylaminopropyl)-2-(trifluoromethyl)phenothiazine monohydrochloride, Adazine, Flumazin, Fluorofen, MC 4703, Neocalm, Neoprin, Nivoman, Siquil hydrochloride, Syquil, Trifluopromazine hydrochloride, Triflupromazine hydrochloride, Triflupromazine monohydrochloride, Vespral, Vesprin, Vesprin hydrochloride, Vetame
InChI Formula InChI=1S/C18H19F3N2S.ClH/c1-22(2)10-5-11-23-14-6-3-4-7-16(14)24-17-9-8-13(12-15(17)23)18(19,20)21;/h3-4,6-9,12H,5,10-11H2,1-2H3;1H
SMILES Cl.CN(C)CCCN1c2ccccc2Sc3ccc(cc13)C(F)(F)F
IUPAC Name N,N-dimethyl-3-[2-(trifluoromethyl)phenothiazin-10-yl]propan-1-amine;hydrochloride
Molecular Weight (g/mol) 388.88
Quantity 250 mg
Formula Weight 388.0988 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.