missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triflupromazine Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal
Specifications
Specifications
| Chemical Name or Material | Triflupromazine hydrochloride |
| CAS | 1098-60-8 |
| Analyte or Component Names | Triflupromazine hydrochloride |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C18 H19 F3 N2 S . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Triflupromazine Hydrochloride, 10H-Phenothiazine-10-propanamine, N,N-dimethyl-2-(trifluoromethyl)-, monohydrochloride (9CI), Phenothiazine, 10-(3-dimethylaminopropyl)-2-(trifluoromethyl)-, hydrochloride (6CI), Phenothiazine, 10-[3-(dimethylamino)propyl]-2-(trifluoromethyl)-, monohydrochloride (8CI), 10-(3-Dimethylaminopropyl)-2-(trifluoromethyl)phenothiazine monohydrochloride, Adazine, Flumazin, Fluorofen, MC 4703, Neocalm, Neoprin, Nivoman, Siquil hydrochloride, Syquil, Trifluopromazine hydrochloride, Triflupromazine hydrochloride, Triflupromazine monohydrochloride, Vespral, Vesprin, Vesprin hydrochloride, Vetame |
| InChI Formula | InChI=1S/C18H19F3N2S.ClH/c1-22(2)10-5-11-23-14-6-3-4-7-16(14)24-17-9-8-13(12-15(17)23)18(19,20)21;/h3-4,6-9,12H,5,10-11H2,1-2H3;1H |
| Solution Type | Single Solution |
| SMILES | Cl.CN(C)CCCN1c2ccccc2Sc3ccc(cc13)C(F)(F)F |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?