missing translation for 'onlineSavingsMsg'
Learn More

Trifluorothymidine, TRC

Product Code. 30453784
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 30453784

Brand: Toronto Research Chemicals TRCT792800500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trifluorothymidine
CAS 70-00-8
Molecular Formula C10 H11 F3 N2 O5
Quantity 500 mg
Synonym Trifluridine, Thymidine','α,α,α-trifluoro-, Uridine','2'-deoxy-5-(trifluoromethyl)- (7CI), α,α,α-Trifluorothymidine, 2'-Deoxy-5-(trifluoromethyl)uridine, 5-(Trifluoromethyl)-2'-deoxyuridine, 5-(Trifluoromethyl)deoxyuridine, 5-Trifluoromethyl-2'-deoxy-β-uridine, 5-Trifluorothymidine, NSC 529182, NSC 75520, Viroptic, 1-(2-Deoxy-β-d-erythro-pentofuranosyl)-5-(trifluoromethyl)pyrimidine-2,4(1H,3H)-dione
InChI Formula InChI=1S/C10H11F3N2O5/c11-10(12,13)4-2-15(9(19)14-8(4)18)7-1-5(17)6(3-16)20-7/h2,5-7,16-17H,1,3H2,(H,14,18,19)/t5-,6+,7+/m0/s1
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N2C=C(C(=O)NC2=O)C(F)(F)F
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-(trifluoromethyl)pyrimidine-2,4-dione
Molecular Weight (g/mol) 296.2
Formula Weight 296.062
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.