Learn More
(Trifluoromethyl)trimethylsilane, 0.5M solution in THF, Thermo Scientific Chemicals
£19.40 - £162.00
Chemical Identifiers
CAS | 81290-20-2, 109-99-9 |
---|---|
Molecular Formula | C5H11F3Si |
Molecular Weight (g/mol) | 156.22 |
MDL Number | MFCD00145454 |
InChI Key | MTYSDPUZDZURPP-UHFFFAOYSA-N |
Synonym | trifluoromethyl trimethylsilane, trimethyl trifluoromethyl silane, trifluoromethyltrimethylsilane, ruppert's reagent, tms-cf3, tfmtms, silane, trimethyl trifluoromethyl, unii-a009786qpj, trimethylsilyl trifluoromethane, ruppert-prakash reagent |
PubChem CID | 552549 |
SMILES | CC(C)(C)[SiH2]C(F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10283002
|
Thermo Scientific Acros
368320050 |
5 mL | Glass bottle |
£19.40
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10374692
|
Thermo Scientific Acros
368320250 |
25 mL | Glass bottle |
£59.20
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10053663
|
Thermo Scientific Acros
368321000 |
100 mL | Glass bottle |
£162.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
81290-20-2, 109-99-9 | |
156.22 | |
MTYSDPUZDZURPP-UHFFFAOYSA-N | |
552549 |
C5H11F3Si | |
MFCD00145454 | |
trifluoromethyl trimethylsilane, trimethyl trifluoromethyl silane, trifluoromethyltrimethylsilane, ruppert's reagent, tms-cf3, tfmtms, silane, trimethyl trifluoromethyl, unii-a009786qpj, trimethylsilyl trifluoromethane, ruppert-prakash reagent | |
CC(C)(C)[SiH2]C(F)(F)F |
Specifications
81290-20-2, 109-99-9 | |
8.74, 92.85 | |
0.8950g/mL | |
-20°C | |
(CH3)3SiCF3 | |
15, 9851 | |
trifluoromethyl trimethylsilane, trimethyl trifluoromethyl silane, trifluoromethyltrimethylsilane, ruppert's reagent, tms-cf3, tfmtms, silane, trimethyl trifluoromethyl, unii-a009786qpj, trimethylsilyl trifluoromethane, ruppert-prakash reagent | |
CC(C)(C)[SiH2]C(F)(F)F | |
156.22 | |
142.2 | |
(Trifluoromethyl)trimethylsilane, 0.5M solution in THF |
7.15, 91.26 | |
Colorless | |
40.0°C | |
C5H11F3Si | |
MFCD00145454 | |
0.895 | |
MTYSDPUZDZURPP-UHFFFAOYSA-N | |
tert-butyl(trifluoromethyl)silane | |
552549 | |
Liquid |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.
May form explosive peroxides.
Suspected of causing cancer.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi
GHS Signal Word: Danger
Recommended Storage : Refrigerator +4°C
- (Trifluoromethyl)trimethylsilane, 0.5M solution in THF
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only