missing translation for 'onlineSavingsMsg'
Learn More

Trifluoromethanesulfonyl chloride, 98%

Product Code. 11420407
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420407 5 g 5g
11430407 25 g 25g
11440407 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420407 Supplier Thermo Scientific Alfa Aesar Supplier No. B21774.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 421-83-0 | CClF3O2S | 168.51 g/mol

Ru(II) complex-catalyzed trifluoromethylating agent for aromatics and alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ru(II) complex-catalyzed trifluoromethylating agent for aromatics and alkenes.

Solubility
Hydrolyzes with water.

Notes
Moisture sensitive. Store away from water/moisture, heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 421-83-0
Molecular Formula CClF3O2S
Molecular Weight (g/mol) 168.51
MDL Number MFCD00007451
InChI Key GRGCWBWNLSTIEN-UHFFFAOYSA-N
Synonym trifluoromethanesulphonyl chloride, triflyl chloride, methanesulfonyl chloride, trifluoro, trifluoromethanesulfonylchloride, trifluoro-methanesulfonyl chloride, methanesulfonylchloride, trifluoro, trifluoromethylsulfonyl chloride, triflic chloride, pfc-msc, cf3so2cl
PubChem CID 79000
IUPAC Name trifluoromethanesulfonyl chloride
SMILES FC(F)(F)S(Cl)(=O)=O

Specifications

Density 1.593
Boiling Point 32°C
Odor Pungent
Refractive Index 1.334
Quantity 5 g
UN Number UN3265
Beilstein 1812016
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes with water.
Formula Weight 168.52
Percent Purity 98%
Chemical Name or Material Trifluoromethanesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.