missing translation for 'onlineSavingsMsg'
Learn More

Trifluoromethanesulfonic Acid Methyl Ester, TRC

Product Code. 30454502
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454502 50 g -
30454070 1 g 1g
30450919 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30454502 Supplier Toronto Research Chemicals Supplier No. TRCT79054050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-27-7
Molecular Formula C2 H3 F3 O3 S
Molecular Weight (g/mol) 164.1
Synonym 1,1,1-Trifluoro-methanesulfonic Acid Methyl Ester
IUPAC Name methyl trifluoromethanesulfonate
SMILES COS(=O)(=O)C(F)(F)F

Specifications

Quantity 50 g
InChI Formula InChI=1S/C2H3F3O3S/c1-8-9(6,7)2(3,4)5/h1H3
Formula Weight 163.9755
Chemical Name or Material Trifluoromethanesulfonic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.