missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic Anhydride, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Trifluoroacetic Anhydride |
| CAS | 407-25-0 |
| Molecular Formula | C4 F6 O3 |
| Quantity | 25 g |
| Synonym | 2,2,2-Trifluoroacetic acid 1,1'-anhydride, 2,2,2-Trifluoroacetic anhydride, Bis(trifluoroacetic) anhydride, Bis(trifluoroacetic acid) anhydride, Hexafluoroacetic anhydride, NSC 96965, Perfluoroacetic anhydride, Trifluoroacetic acid anhydride, Trifluoroacetic anhydride, Trifluoroacetyl anhydride, TFAA |
| InChI Formula | InChI=1S/C4F6O3/c5-3(6,7)1(11)13-2(12)4(8,9)10 |
| SMILES | FC(F)(F)C(=O)OC(=O)C(F)(F)F |
| IUPAC Name | (2,2,2-trifluoroacetyl) 2,2,2-trifluoroacetate |
| Molecular Weight (g/mol) | 210.03 |
| Formula Weight | 209.9752 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction