missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic acid in Water, 0.1% (v/v), BAKER ANALYZED™, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
:
2.5 L
Description
- For Laboratory, Research, or Manufacturing Use
- Filtered through a 0.2 micron filter.
- Trifluoroacetic Acid: 0.095 - 0.105 %
- Gradient Elution Test (a.u.) - 254 nm: ≤ 0.01
- LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine): ≤ 50 ppb
Specifications
Specifications
| Appearance | Passes Test |
| Synonym | TFA |
| Quantity | 2.5 L |
| Packaging | Glass bottle |
| Residue after Evaporation | ≤1.0 ppm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction