missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic Acid, Thermo Scientific

Product Code. 30280960 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
2 mL
5 mL
50 mL
10 x 1 mL
Unit Size:
1mL
2mL
50mL
5mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280960 50 mL 50mL
30280970 1 mL 1mL
30280966 2 mL 2mL
30280947 5 mL 5mL
30280948 10 x 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30280960 Supplier Thermo Scientific Supplier No. T001160050
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
  • Ideal high purity mobile phase additive for LC/MS and UHPLC-UV applications
  • Evaluated for metal impurities at ppb concentrations for minimal metal mass adduct formation
  • Screened for UV-absorbing contaminants to reduce interferences and for smoother baselines
  • TFA is a common mobile phase additive that can function as an ion pair reagent
TRUSTED_SUSTAINABILITY

Specifications

Item Name Trifluoroacetic Acid (TFA)
Form Liquid
Reagent Type Ion Pairing Reagent
Quantity 50 mL
Formulation 99.5% TFA
For Use With LC/MS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.