missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, silver salt, 98%

Product Code. 10041550 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10041550 5 g Glass bottle 5g
10615002 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10041550 Supplier Thermo Scientific Acros Supplier No. 139730050
 Restricted Item
Request Bulk or Custom Formats

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 2966-50-9 | C2AgF3O2 | 220.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2966-50-9
Molecular Formula C2AgF3O2
Molecular Weight (g/mol) 220.89
MDL Number MFCD00013199
InChI Key KZJPVUDYAMEDRM-UHFFFAOYSA-M
Synonym silver trifluoroacetate, silver 1+ trifluoroacetate, silver i 2,2,2-trifluoroacetate, silver mono trifluoroacetate, trifluoroacetic acid silver salt, silver 1+ ion trifluoroacetate, acetic acid, trifluoro-, silver 1+ salt, silver i trifluoroacetate, trifluoroacetic acid, silver salt, trifluoroacetic acid, silver 1+ salt
PubChem CID 76299
IUPAC Name silver;2,2,2-trifluoroacetate
SMILES C(=O)(C(F)(F)F)[O-].[Ag+]

Specifications

Melting Point 257.0°C to 260.0°C
Color Beige to Gray
Infrared Spectrum Authentic
Assay Percent Range 0.98
Packaging Glass bottle
Linear Formula CF3CO2Ag
Quantity 5 g
Fieser 01,1018; 07,323; 08,444; 11,471; 16,301; 17,154
Solubility Information Solubility in water: soluble
Formula Weight 220.89
Percent Purity 0.98
Physical Form Crystalline Powder
Chemical Name or Material Trifluoroacetic acid, silver salt, 0.98
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetic acid, silver salt
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.