missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic acid, Reagent Grade, 99%, Honeywell™
Reagent Grade, 99%
£14.74 - £652.99
Chemical Identifiers
CAS | 76-05-1 |
---|---|
Molecular Formula | C2HF3O2 |
Molecular Weight (g/mol) | 114.02 |
MDL Number | MFCD00004169 |
InChI Key | DTQVDTLACAAQTR-UHFFFAOYSA-N |
Synonym | trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova |
PubChem CID | 6422 |
ChEBI | CHEBI:45892 |
IUPAC Name | trifluoroacetic acid |
SMILES | OC(=O)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15698490
|
Honeywell Research Chemicals
T6508-5ML |
5 mL | |||||||
15688490
|
Honeywell Research Chemicals
T6508-25ML |
25 mL | |||||||
15668190
|
Honeywell Research Chemicals
T6508-100ML |
100 mL | |||||||
15628200
|
Honeywell Research Chemicals
T6508-500ML |
500 mL | |||||||
15608200
|
Honeywell Research Chemicals
T6508-1L |
1 L | |||||||
15618200
|
Honeywell Research Chemicals
T6508-2L |
2 L | |||||||
15698190
|
Honeywell Research Chemicals
T6508-1AMP |
1 Ampule | |||||||
15678190
|
Honeywell Research Chemicals
T6508-10AMP |
10 Ampules |
£71.41
10 ampoules |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15638200
|
Honeywell Research Chemicals
T6508-5X10AMP |
5 x 10 Ampules |
£328.49
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15688190
|
Honeywell Research Chemicals
T6508-10X10AMP |
10 x 10 Ampules | |||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
76-05-1 | |
114.02 | |
DTQVDTLACAAQTR-UHFFFAOYSA-N | |
6422 | |
trifluoroacetic acid |
C2HF3O2 | |
MFCD00004169 | |
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
CHEBI:45892 | |
OC(=O)C(F)(F)F |
Specifications
76-05-1 | |
CF3COOH | |
UN2699 | |
trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova | |
OC(=O)C(F)(F)F | |
114.02 | |
CHEBI:45892 | |
99% | |
1.489g/mL at 20°C | |
3.9 (vs air) | |
Trifluoroacetic acid |
C2HF3O2 | |
MFCD00004169 | |
742035 | |
DTQVDTLACAAQTR-UHFFFAOYSA-N | |
trifluoroacetic acid | |
6422 | |
114.02g/mol | |
ReagentPlus™ | |
>100°C (>212°F) | |
97.5 mmHg (20°C) |
Safety and Handling
P261-P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
missing translation for 'rtecsNumber' : AJ9625000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Trifluoroacetic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.