missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, HPLC Grade, 99.5+%

Product Code. 11455440
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 x 100 mL
4 x 500 mL
Unit Size:
100mL
2000mL
400mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455440 4 x 100 mL 400mL
11465440 100 mL 100mL
11475440 500 mL 500mL
11485440 4 x 500 mL 2000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11455440 Supplier Thermo Scientific Alfa Aesar Supplier No. 044630.5Y
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 76-05-1 | C2HF3O2 | 114.02 g/mol

The combination of properties like solubility in most of the solvents, volatility, catalytic property and strong acidity with non-oxidizing nature makes it a widely used reagent in an organic synthesis. Trifluoroacetic acid is an important building block in the synthesis of pharmaceuticals, agrochemicals and ceramic compounds. It is a precursor to many fluorinated compounds, and widely used in peptide synthesis and other organic transformations involving deprotection of t-BOC group. It finds use as an ion pairing agent in liquid chromatography, as a solvent in NMR spectroscopy and as a calibrant in mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name 2,2,2-trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Melting Point -15°C
Boiling Point 72°C to 73°C
Color Colorless
Assay Percent Range >99.5%
UN Number 2699
Beilstein 742035
Solubility Information Miscible with water,ether,acetone,ethanol,benzene,hexane and carbon tetrachloride.
Formula Weight 114.02
Grade HPLC
Density 1.48 g/cm3 at 20°C
Quantity 4 x 100 mL
Odor Pungent
Physical Form Liquid
Refractive Index 1.284
Sensitivity Hygroscopic
Vapor Pressure 8mm Hg at 20°C
Chemical Name or Material Trifluoroacetic Acid, Water 0.1%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.