missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, Eluent additive for LC-MS, Solstice

Product Code. 15665440 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
10 mL
5 x 10 mL
1 L
Packaging:
Ampoule
Glass bottle
Unit Size:
1 L
10mL
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15665440 5 x 10 mL Ampoule Pack of 5
15618400 10 mL Ampoule 10mL
15655440 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15665440 Supplier Honeywell-Fluka Supplier No. 409675X10ML
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Eluent additive for LC-MS

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
SMILES OC(=O)C(F)(F)F

Specifications

Absorbance max. 0.025 at 320 nm, max. 0.03 at 300 nm, max. 0.04 at 290 nm, max. 0.05 280 nm, max. 0.1 at 270 nm, max. 0.9 at 260 nm
Melting Point -15.4°C (literature)
Boiling Point 72°C
CAS Max % 100.0000%
Linear Formula CF3COOH
UN Number UN2699
Beilstein 742035
IUPAC Name 2,2,2-trifluoroacetic acid
Formula Weight 114.02
Percent Purity >99%
Grade LC-MS
Density 1.489 g/mL at 20 °C(lit.)
Quantity 5 x 10 mL
Flash Point 100°C
Packaging Ampoule
Vapor Density 3.9 (vs air)
Vapor Pressure 97.5 mmHg (20°C)
Chemical Name or Material Trifluoroacetic acid, Eluent additive for LC-MS
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.