missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, 99%, Solstice

Product Code. 15638400 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
10 x 1 mL
Packaging:
Ampoule
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15638400 10 x 1 mL Ampoule Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15638400 Supplier Honeywell Fluka Supplier No. 4096710X1ML
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Eluent additive for liquid chromatography-mass spectrometry (LC-MS) packaged in ten 1mL ampoules.

TRUSTED_SUSTAINABILITY

Specifications

Absorbance Max. 0.025 at 320nm, Max. 0.035 at 300nm, Max. 0.04 at 290nm, Max. 0.05 at 280nm, Max. 0.1 at 270nm
CAS 76-05-1
Melting Point −15°C
Boiling Point 72°C
Molecular Formula C2HF3O2
Linear Formula CF3COOH
MDL Number MFCD00004169
UN Number 2699
Beilstein 742035
Synonym TFA
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
SMILES OC(=O)C(F)(F)F
IUPAC Name trifluoroacetic acid
Molecular Weight (g/mol) 114.02
Percent Purity 99.0% (min.)
Density 1.48g/cm3
Quantity 10 x 1 mL
Flash Point 100°C
Packaging Ampoule
Refractive Index n20/D 1.3
Vapor Density 3.9 vs. air
Vapor Pressure 97.5mm Hg at 20°C
Chemical Name or Material Trifluoroacetic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.