missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetamidine, tech. 85%

Product Code. 11409830
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409830 100 g 100g
11328795 5 g 5g
11338795 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409830 Supplier Thermo Scientific Alfa Aesar Supplier No. L12422.22
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 354-37-0 | C2H3F3N2 | 112.06 g/mol

Trifluoroacetamidine is used as probe for determination of membrane potential and extra/intracellular volume of erythrocytes by fluorine-19 NMR studies. It is a reagent used as protected ammonia equivalent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trifluoroacetamidine is used as probe for determination of membrane potential and extra/intracellular volume of erythrocytes by fluorine-19 NMR studies. It is a reagent used as protected ammonia equivalent.

Solubility
Fully miscible in water.

Notes
Store at -20°C. Incompatible with oxidizing agents. Moisture Sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 354-37-0
Molecular Formula C2H3F3N2
Molecular Weight (g/mol) 112.06
MDL Number MFCD00041545
InChI Key NITMACBPVVUGOJ-UHFFFAOYSA-N
Synonym trifluoroacetamidine, trifluoroethanimidamide, 2,2,2-trifluoroacetamidine, ethanimidamide, 2,2,2-trifluoro, sntljqhjptadtaetp@, trifluoroacetoamidine, trifluoroacetamideine, trifluoro-acetamidine, acmc-20aje4, 2,2,2-trifluoroacetoamidine
PubChem CID 2776882
IUPAC Name 2,2,2-trifluoroethanimidamide
SMILES NC(=N)C(F)(F)F

Specifications

Density 1.45
Boiling Point 35°C to 37°C (11 mmHg)
Refractive Index 1.382
Quantity 100 g
UN Number UN3267
Beilstein 605978
Sensitivity Moisture sensitive
Solubility Information Fully miscible in water.
Formula Weight 112.05
Percent Purity 85%
Grade Technical
Chemical Name or Material Trifluoroacetamidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.