missing translation for 'onlineSavingsMsg'
Learn More
Learn More


Trifluoroacetaldehyde ethyl hemiacetal, 90%, Thermo Scientific™
Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Density | 1.2400g/mL |
Color | Colorless |
Boiling Point | 104°C to 105°C (745.0mmHg) |
Flash Point | 39°C |
Assay Percent Range | 88% min. (GC) |
Infrared Spectrum | Authentic |
Linear Formula | CF3CH(OC2H5)OH |
Beilstein | 01, IV, 3133 |
Packaging | Glass bottle |
Refractive Index | 1.3410 to 1.344 |
Show More |
Safety and Handling


Product Identifier
- Trifluoroacetaldehyde ethyl hemiacetal
- Danger
- Acute toxicity Category 4
- Flammable liquid Category 2
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only