Learn More
Trifluoroacetaldehyde ethyl hemiacetal, 90%, Acros Organics
Brand: Acros Organics 139710050
Code : 29
Additional Details : CAS Number : 433-27-2 Weight : 0.00600kg Transport : UN number : 1989 Chem class : 3 Pack group : III
Packaging | Glass bottle |
---|---|
Quantity | 5mL |
Chemical Identifiers
433-27-2 | |
144.09 | |
KLXJPQNHFFMLIG-UHFFFAOYSA-N | |
9897 | |
CCOC(C(F)(F)F)O |
C4H7F3O2 | |
MFCD00000441 | |
trifluoroacetaldehyde ethyl hemiacetal, fluoral ethyl hemiacetal, ethanol, 1-ethoxy-2,2,2-trifluoro, trifluoroacetaldehyde hemiethylacetal, 1-ethoxy-2,2,2-trifluoro-ethanol, trifluoroacetaldehyde ethylhemiacetal, 1-ethoxy-2,2,2-trifluoroethan-1-ol, pubchem12643, acmc-1aj38, 1-ethoxy-2,2-trifluoroethanol | |
1-ethoxy-2,2,2-trifluoroethanol |
Specifications
Trifluoroacetaldehyde ethyl hemiacetal | |
39°C | |
Glass bottle | |
1.24 | |
Colorless | |
90% | |
88% min. (GC) | |
CF3CH(OC2H5)OH | |
01, IV, 3133 | |
KLXJPQNHFFMLIG-UHFFFAOYSA-N | |
1-ethoxy-2,2,2-trifluoroethanol | |
9897 | |
Liquid |
1.2400g/mL | |
Authentic | |
1.3410 to 1.344 | |
104°C to 105°C (745.0mmHg) | |
5mL | |
433-27-2 | |
C4H7F3O2 | |
MFCD00000441 | |
trifluoroacetaldehyde ethyl hemiacetal, fluoral ethyl hemiacetal, ethanol, 1-ethoxy-2,2,2-trifluoro, trifluoroacetaldehyde hemiethylacetal, 1-ethoxy-2,2,2-trifluoro-ethanol, trifluoroacetaldehyde ethylhemiacetal, 1-ethoxy-2,2,2-trifluoroethan-1-ol, pubchem12643, acmc-1aj38, 1-ethoxy-2,2-trifluoroethanol | |
CCOC(C(F)(F)F)O | |
144.09 | |
144.09 | |
90% |
Safety and Handling
- Trifluoroacetaldehyde ethyl hemiacetal
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.