missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoperazine.2HCl, Lipomed
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Trifluoperazine.2HCl
Specifications
Specifications
| Chemical Name or Material | Trifluoperazine hydrochloride |
| CAS | 440-17-5 |
| Analyte or Component Names | Trifluoperazine hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H24 F3 N3 S . 2 Cl H |
| Synonym | Trifluoperazine hydrochloride, 10-[3-(4-methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)-10 H-phenothiazine dihydrochloride, 10 H-Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-2-(trifluoromethyl)-, hydrochloride (1:2)10 H-Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-2-(trifluoromethyl)-, dihydrochloride (9 CI), Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-2-(trifluoromethyl)-, dihydrochloride (6 CI,8 CI), 2-Trifluoromethyl-10-[3-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride, 2-Trifluoromethyl-10-[3'-(1-methyl-4-piperazinyl)propyl]phenothiazine dihydrochloride |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.Cl.CN1CCN(CCCN2c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1 |
| IUPAC Name | 10-[3-(4-methylpiperazin-1-yl)propyl]-2-(trifluoromethyl)phenothiazine;dihydrochloride |
| Industry Type | Pharmaceutical |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction