missing translation for 'onlineSavingsMsg'
Learn More

Triflumuron, 98+%, Thermo Scientific Chemicals

Product Code. 15405389
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15405389 100 g 100g
15485379 10 g 10g
15495379 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15405389 Supplier Thermo Scientific Alfa Aesar Supplier No. J63112.22
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Triflumuron acts as a chitin synthesis inhibitor and prevents uridine incorporation into RNA

Triflumuron acts as a chitin synthesis inhibitor and prevents uridine incorporation into RNA

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64628-44-0
Molecular Formula C15H10ClF3N2O3
Molecular Weight (g/mol) 358.701
MDL Number MFCD00072496
InChI Key XAIPTRIXGHTTNT-UHFFFAOYSA-N
Synonym triflumuron, alsystin, trifluron, mascot, alsystine, triflumuron iso, caswell no. 217c, bay-sir 8514, unii-3ft64dyg8k, bay-vi 7533
PubChem CID 47445
ChEBI CHEBI:39388
IUPAC Name 2-chloro-N-[[4-(trifluoromethoxy)phenyl]carbamoyl]benzamide
SMILES C1=CC=C(C(=C1)C(=O)NC(=O)NC2=CC=C(C=C2)OC(F)(F)F)Cl

Specifications

Melting Point 198°C
Quantity 100 g
Beilstein 2776684
Merck Index 14,9678
Solubility Information Soluble in DMF
Formula Weight 358.7
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material Triflumuron

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.