missing translation for 'onlineSavingsMsg'
Learn More

Trifarotene, MedChemExpress

Product Code. p-200102117
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16492878

Brand: Medchem Express HY100256

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Trifarotene (CD5789) is a potent and selective RARγ agonist. Trifarotene (CD5789) shows ∼65-fold and ∼16-fold selectivitiy for the RARγ (EC50=7.7 nM) over RARα (EC50=500 nM) and RARβ (EC50=125 nM), respectively.

Trifarotene (CD5789) is a potent and selective RARγ agonist. Trifarotene (CD5789) shows ∼65-fold and ∼16-fold selectivitiy for the RARγ (EC50=7.7 nM) over RARα (EC50=500 nM) and RARβ (EC50=125 nM), respectively.

Specifications

Chemical Name or Material Trifarotene
CAS 895542-09-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H33NO4
Quantity 10 mg
Synonym CD5789
Solubility Information DMSO : 250 mg/mL (543.97 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C2=CC=C(OCCO)C(C3=CC=C(N4CCCC4)C(C(C)(C)C)=C3)=C2)C=C1)O
Molecular Weight (g/mol) 459.58
Formula Weight 459.58
Percent Purity 99.5%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.