missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIF/TICAM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-31189SS
This item is not returnable.
View return policy
Description
TRIF/TICAM1 Polyclonal specifically detects TRIF/TICAM1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| TRIF/TICAM1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| TICAM1 | |
| Antibody was raised against a peptide corresponding to 14 amino acids near the C-terminus of human TRIF. The immunogen is located within the last 50 amino acids of TRIF. Amino Acid Squence: RGSQAPEDKTQEAE | |
| Ion exchange chromatography | |
| Signal Transduction | |
| 148022 | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry 10 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/mL, Immunohistochemistry-Paraffin 10 ug/ml, Immunofluorescence 10 ug/ml | |
| MGC35334, Proline-rich, vinculin and TIR domain-containing protein B, PRVTIRBTIR domain-containing adapter protein inducing IFN-beta, Putative NF-kappa-B-activating protein 502H, TICAM-1TIR domain containing adaptor inducing interferon-beta, TIR domain-containing adapter molecule 1, TIR domain-containing adaptor molecule 1, toll-like receptor adaptor molecule 1, TRIFToll-interleukin-1 receptor domain-containing adapter protein inducinginterferon beta | |
| Rabbit | |
| 55 kDa | |
| 0.025 mg | |
| Primary | |
| Antibody is specific for human TRIF. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction