missing translation for 'onlineSavingsMsg'
Learn More

TRIF/TICAM1 Antibody, Novus Biologicals™

Product Code. 18387782 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18387782 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18387782 Supplier Novus Biologicals Supplier No. NBP312427

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TRIF/TICAM1 Polyclonal antibody specifically detects TRIF/TICAM1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIF/TICAM1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:750, ELISA 1:20000
Formulation Contains Tris, HCl/glycine buffer (pH 7.4-7.8), 30% glycerol, 0.5% BSA, along with cryo-protective agents, and HEPES (exact storage and stabilization buffer information is proprietary) with 0.02% Sodium Azide
Gene Alias MGC35334, Proline-rich, vinculin and TIR domain-containing protein B, PRVTIRBTIR domain-containing adapter protein inducing IFN-beta, Putative NF-kappa-B-activating protein 502H, TICAM-1TIR domain containing adaptor inducing interferon-beta, TIR domain-containing adapter molecule 1, TIR domain-containing adaptor molecule 1, toll-like receptor adaptor molecule 1, TRIFToll-interleukin-1 receptor domain-containing adapter protein inducinginterferon beta
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 150-199 on human E3 ubiquitin-protein ligase RNF138 protein.
Purification Method Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 148022
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.