missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIF/TICAM1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB120-13810G
This item is not returnable.
View return policy
Description
TRIF/TICAM1 Polyclonal specifically detects TRIF/TICAM1 in Mouse, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, Knockdown Validated.
Specifications
| TRIF/TICAM1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunoblotting, Knockdown Validated | |
| MGC35334, Proline-rich, vinculin and TIR domain-containing protein B, PRVTIRBTIR domain-containing adapter protein inducing IFN-beta, Putative NF-kappa-B-activating protein 502H, TICAM-1TIR domain containing adaptor inducing interferon-beta, TIR domain-containing adapter molecule 1, TIR domain-containing adaptor molecule 1, toll-like receptor adaptor molecule 1, TRIFToll-interleukin-1 receptor domain-containing adapter protein inducinginterferon beta | |
| Synthetic peptide made to an internal portion of the mouse TRIF protein (between amino acids 100-180) [UniProt Q80UF7]. | |
| 0.1 mL | |
| Signal Transduction | |
| 148022 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoblot, KnockDown | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat, Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction