missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRIF Recombinant Rabbit Monoclonal Antibody (PSH05-39)

Description
TRIF Recombinant Monoclonal Antibody for Western Blot
Involved in innate immunity against invading pathogens. Adapter used by TLR3 and TLR4 (through TICAM2) to mediate NF-kappa-B and interferon-regulatory factor (IRF) activation, and to induce apoptosis. Ligand binding to these receptors results in TRIF recruitment through its TIR domain. Distinct protein-interaction motifs allow recruitment of the effector proteins TBK1, TRAF6 and RIPK1, which in turn, lead to the activation of transcription factors IRF3 and IRF7, NF-kappa-B and FADD respectively.
Specifications
Specifications
| Antigen | TRIF |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | PSH05-39 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | TICAM1 |
| Gene Accession No. | Q8IUC6 |
| Gene Alias | AW046014; AW547018; hCG_39321; HSD34; IIAE6; MyD88-3; Proline-rich, vinculin and TIR domain-containing protein B; PRVTIRB; Putative NF-kappa-B-activating protein 502H; RNF36; TI; Ticam1; TICAM-1; TIR domain containing adaptor inducing interferon-beta; TIR domain-containing adapter molecule 1; TIR domain-containing adapter protein inducing IFN-beta; TIR domain-containing adaptor inducing interferon-beta; TIR-domain containing adaptor inducing interferon-beta; toll like receptor adaptor molecule 1; toll-interleukin 1 receptor domain-containing adapter protein inducing interferon-beta; toll-interleukin I receptor domain containing adaptor molecule 1; toll-interleukin-1 receptor domain-containing adapter protein inducing interferon beta; toll-like receptor adaptor molecule 1; TRIF |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?