missing translation for 'onlineSavingsMsg'
Learn More

Triethyltin chloride, Thermo Scientific Chemicals

Product Code. 11334527
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334527 5 g 5g
11344527 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334527 Supplier Thermo Scientific Alfa Aesar Supplier No. 039601.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 994-31-0 | Molecular Formula: C6H17ClSn | Molecular Weight: 243.36 g/mol

It is used in the synthesis of some tin alkyls and also in the preparation of organolithium compounds by the transmetalation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 994-31-0
Molecular Formula C6H17ClSn
Molecular Weight (g/mol) 243.36
MDL Number MFCD00040414
InChI Key PFMCXCNQGDRWRV-UHFFFAOYSA-N
Synonym triethyltin chloride, stannane, chlorotriethyl, chlorotriethyltin, triethylchlorotin, triethylchlorostannane, triethylstannyl chloride, tin, triethyl-, chloride, triethyltin chloride c2h5 3sncl, triethylstannium chloride, triaethylzinnchlorid
PubChem CID 13820
IUPAC Name chloro(triethyl)stannane
SMILES Cl.CC[SnH](CC)CC

Specifications

Density 1.429g/mL
Boiling Point 206°C
Odor Unpleasant
Quantity 5 g
UN Number UN2788
Sensitivity Moisture Sensitive
Formula Weight 241.33
Physical Form Liquid
Chemical Name or Material Triethyltin chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.