missing translation for 'onlineSavingsMsg'
Learn More

Triethyltin bromide, Thermo Scientific™

Product Code. 11384819
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11384819 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11384819 Supplier Thermo Scientific Alfa Aesar Supplier No. 071135.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2767-54-6
Molecular Formula C6H17BrSn
Molecular Weight (g/mol) 287.82
MDL Number MFCD00000052
InChI Key MXGZXTQIURJKMF-UHFFFAOYSA-N
Synonym triethyltin bromide, stannane, bromotriethyl, triethyl tin bromide, triethylstannium bromide, tin, triethyl-, bromide, unii-uop17166kc, triethylstannyl bromide, bromo triethyl stannane, stannane,bromotriethyl, acmc-1cl7y
PubChem CID 17701
IUPAC Name bromo(triethyl)stannane
SMILES Br.CC[SnH](CC)CC

Specifications

Melting Point -13.5°C
Density 1.63 g/mL
Boiling Point 223°C to 225°C
Flash Point 99°C (210°F)
Refractive Index 1.52
Quantity 1 g
UN Number UN2788
Sensitivity Moisture sensitive
Formula Weight 285.79
Physical Form Liquid
Chemical Name or Material Triethyltin bromide
Show More Show Less
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.