missing translation for 'onlineSavingsMsg'
Learn More

Triethylsulfonium bis(trifluoromethylsulfonyl)imide, 98%

Product Code. 11466210
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466210 50 g 50g
11456210 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11466210 Supplier Thermo Scientific Alfa Aesar Supplier No. H27116.18
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 321746-49-0 | C8H15F6NO4S3 | 399.381 g/mol

Proton-Conducting Properties of a Bronsted Acid-Base Ionic Liquid and Ionic Melts Consisting of Bis(trifluoromethanesulfonyl)imide and Benzimidazole for Fuel Cell Electrolytes. Symmetric and aliphatic trialkylsulfonium cation form room temperature molten salts (RTMS) with bis(trifluoromethylsulfonyl)imide (TFSI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Proton-Conducting Properties of a Bronsted Acid-Base Ionic Liquid and Ionic Melts Consisting of Bis(trifluoromethanesulfonyl)imide and Benzimidazole for Fuel Cell Electrolytes. Symmetric and aliphatic trialkylsulfonium cation form room temperature molten salts (RTMS) with bis(trifluoromethylsulfonyl)imide (TFSI).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 321746-49-0
Molecular Formula C8H15F6NO4S3
Molecular Weight (g/mol) 399.381
MDL Number MFCD08276438
InChI Key BLODSRKENWXTLO-UHFFFAOYSA-N
Synonym triethylsulfonium bis trifluoromethylsulfonyl imide, triethylsulfonium bis trifluoromethanesulfonyl imide, set3 ntf2, dsstox_cid_27864, dsstox_rid_82617, dsstox_gsid_47888, bistriflylimide anion; triethylsulfanium, triethylsulfonium bis trifluoromethylsulfonyl imide, for electrochemistry qnmr
PubChem CID 16211135
IUPAC Name bis(trifluoromethylsulfonyl)azanide;triethylsulfanium
SMILES CC[S+](CC)CC.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Quantity 50 g
Formula Weight 399.39
Percent Purity 98%
Chemical Name or Material Triethylsulfonium bis(trifluoromethylsulfonyl)imide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.