missing translation for 'onlineSavingsMsg'
Learn More

Triethylsilyl trifluoromethanesulfonate, 98%

Product Code. 11303546
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11303546 5 g 5g
11313546 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11303546 Supplier Thermo Scientific Alfa Aesar Supplier No. L14477.06
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 79271-56-0 | C7H15F3O3SSi | 264.33 g/mol

Triethylsilyl trifluoromethanesulfonate acts as a silylating agent. It is also useful as a Lewis acid catalyst. Further, it reacts with 1-diazo-3,3-dimethyl-butan-2-one to prepare 1-diazo-3,3-dimethyl-1-(triethylsilyl)-2-butanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylsilyl trifluoromethanesulfonate acts as a silylating agent. It is also useful as a Lewis acid catalyst. Further, it reacts with 1-diazo-3,3-dimethyl-butan-2-one to prepare 1-diazo-3,3-dimethyl-1-(triethylsilyl)-2-butanone.

Solubility
Miscible with dichloromethane, hydrocarbons, dialkyl ethers and halogenated solvents. Immiscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79271-56-0
Molecular Formula C7H15F3O3SSi
Molecular Weight (g/mol) 264.33
MDL Number MFCD00000407
InChI Key STMPXDBGVJZCEX-UHFFFAOYSA-N
Synonym triethylsilyl triflate, triethylsilyl trifluoromethanesulphonate, tes triflate, triethylsilyltriflate, triethylsilyltrifluoromethanesulfonate, trifluoromethanesulfonic acid triethylsilyl ester, 1,1,1-triethylsilyl trifluoromethanesulfonate, methanesulfonic acid, trifluoro-, triethylsilyl ester, et3siotf, tesotf
PubChem CID 2733308
IUPAC Name triethylsilyl trifluoromethanesulfonate
SMILES CC[Si](CC)(CC)OS(=O)(=O)C(F)(F)F

Specifications

Density 1.225
Boiling Point 207°C
Flash Point 72°C (161°F)
Linear Formula CF3SO3Si(CH2CH3)3
Refractive Index 1.391
Quantity 5 g
UN Number UN3265
Beilstein 3590541
Sensitivity Moisture sensitive
Solubility Information Miscible with dichloromethane,hydrocarbons,dialkyl ethers and halogenated solvents. Immiscible with water.
Formula Weight 264.33
Percent Purity 98%
Chemical Name or Material Triethylsilyl trifluoromethanesulfonate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.