missing translation for 'onlineSavingsMsg'
Learn More

Triethylsilanol, 97%

Product Code. 11332286
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332286 5 g 5g
11322286 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332286 Supplier Thermo Scientific Alfa Aesar Supplier No. L03738.06
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 597-52-4 | C6H16OSi | 132.278 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 597-52-4
Molecular Formula C6H16OSi
Molecular Weight (g/mol) 132.278
MDL Number MFCD00042640
InChI Key WVMSIBFANXCZKT-UHFFFAOYSA-N
Synonym triethylsilanol, silanol, triethyl, hydroxytriethylsilane, silanol, 1,1,1-triethyl, triethyl hydroxy silane, triethyl hydroxy silicon, tesoh, c2h5 3sioh, acmc-1an47
PubChem CID 69005
IUPAC Name triethyl(hydroxy)silane
SMILES CC[Si](CC)(CC)O

Specifications

Boiling Point 154°C
Quantity 5 g
UN Number UN1993
Beilstein 1732848
Flash Point 57°C (135°F)
Formula Weight 132.28
Percent Purity 97%
Odor Mild
Refractive Index 1.433
Density 0.86
Chemical Name or Material Triethylsilanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.