missing translation for 'onlineSavingsMsg'
Learn More

Triethylphosphine oxide

Product Code. 11307268
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11307268 1 g 1g
11317268 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11307268 Supplier Thermo Scientific Alfa Aesar Supplier No. 030391.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 597-50-2 | C6H15OP | 134.16 g/mol

Triethylphosphine oxide is used as a probe molecule and systems are evaluated by 31P NMR spectroscopy. Triethylphosphine oxide is used to produce tris(perfluoroethyl)difluorophosphane. This reaction will need reagent of HF. This reaction needs electrochemical fluorination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 597-50-2
Molecular Formula C6H15OP
Molecular Weight (g/mol) 134.16
MDL Number MFCD00013914
InChI Key ZSSWXNPRLJLCDU-UHFFFAOYSA-N
Synonym triethylphosphine oxide, phosphine oxide, triethyl, triethylphoshorus oxide, c2h5 3p=o, triethylphospine oxide, et3po, triethylphosphino-1-one, triethyl oxo phosphorane, diethylphosphoryl ethane, triethylphosphine oxide 1g
PubChem CID 79061
IUPAC Name 1-diethylphosphorylethane
SMILES CCP(=O)(CC)CC

Specifications

Melting Point 48°C to 50°C
Color White
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 134.16
Physical Form Crystalline
Chemical Name or Material Triethylphosphine oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.