missing translation for 'onlineSavingsMsg'
Learn More

Triethylphosphine oxide, 98+%, Thermo Scientific™

Product Code. 10167582 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10167582 1 g Glass bottle 1g
10649384 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10167582 Supplier Thermo Scientific Acros Supplier No. 380030010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 597-50-2
Molecular Formula C6H15OP
Molecular Weight (g/mol) 134.16
MDL Number MFCD00013914
InChI Key ZSSWXNPRLJLCDU-UHFFFAOYSA-N
Synonym triethylphosphine oxide, phosphine oxide, triethyl, triethylphoshorus oxide, c2h5 3p=o, triethylphospine oxide, et3po, triethylphosphino-1-one, triethyl oxo phosphorane, diethylphosphoryl ethane, triethylphosphine oxide 1g
PubChem CID 79061
IUPAC Name (diethylphosphoryl)ethane
SMILES CCP(=O)(CC)CC

Specifications

Melting Point 52.0°C to 53.0°C
Color White
Boiling Point 84.0°C to 85.0°C (3.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 1 g
Formula Weight 134.16
Percent Purity ≥98%
Physical Form Needles
Chemical Name or Material Triethylphosphine oxide
Show More Show Less
compliance-icons
Product Identifier
  • Triethylphosphine oxide
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.