missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethyloxonium tetrafluoroborate, 1.0M in dichloromethane, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Description
Triethyloxonium tetrafluoroborate is used as an effective agent used for ethylation., laboratory chemicals, manufacture of substances.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Triethyloxonium tetrafluoroborate is used as an effective agent used for ethylation., laboratory chemicals, manufacture of substances.
Solubility
Soluble in Dichloromethane. Reacts with water.
Notes
Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Triethyloxonium tetrafluoroborate is used as an effective agent used for ethylation., laboratory chemicals, manufacture of substances.
Solubility
Soluble in Dichloromethane. Reacts with water.
Notes
Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Density | 1.328 |
Odor | Chloroform-like |
Linear Formula | (CH3CH2)3OBF4 |
Quantity | 25 mL |
UN Number | UN2922 |
Beilstein | 3598090 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in Dichloromethane. Reacts with water. |
Formula Weight | 189.99 |
Concentration or Composition (by Analyte or Components) | 1.0M in dichloromethane |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'