missing translation for 'onlineSavingsMsg'
Learn More

Triethyloxonium tetrafluoroborate, 1.0M in dichloromethane

Product Code. 11493910
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493910 100 mL 100mL
11483910 25 mL 25mL
11403920 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493910 Supplier Thermo Scientific Alfa Aesar Supplier No. A11678.AE
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 368-39-8 | C6H15BF4O | 189.99 g/mol

Triethyloxonium tetrafluoroborate is used as an effective agent used for ethylation., laboratory chemicals, manufacture of substances.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyloxonium tetrafluoroborate is used as an effective agent used for ethylation., laboratory chemicals, manufacture of substances.

Solubility
Soluble in Dichloromethane. Reacts with water.

Notes
Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 368-39-8
Molecular Formula C6H15BF4O
Molecular Weight (g/mol) 189.99
MDL Number MFCD00044423
InChI Key IYDQMLLDOVRSJJ-UHFFFAOYSA-N
Synonym triethyloxonium tetrafluoroborate, triethyloxonium fluoroborate, triethyloxonium fluoborate, triethyloxonium borofluoride, triethoxonium fluoroborate, meerwein's reagent, triethyloxidanium tetrafluoroborate, unii-z0b19dd36j, triethyloxonium tetraflouroborate, boron tetrafluoride triethyl oxonium
PubChem CID 2723982
IUPAC Name triethyloxidanium;tetrafluoroborate
SMILES F[B-](F)(F)F.CC[O+](CC)CC

Specifications

Density 1.328
Odor Chloroform-like
Linear Formula (CH3CH2)3OBF4
Quantity 100 mL
UN Number UN2922
Beilstein 3598090
Sensitivity Moisture sensitive
Solubility Information Soluble in Dichloromethane. Reacts with water.
Formula Weight 189.99
Concentration or Composition (by Analyte or Components) 1.0M in dichloromethane
Chemical Name or Material Triethyloxonium tetrafluoroborate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.