missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethylgermanium hydride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Co-reactant for the preparation of germanium acrylates. They are the Synthetic Reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 1.004 g/mL |
Boiling Point | 122°C |
Flash Point | 5°C (41°F) |
Refractive Index | 1.4382 |
Quantity | 1 g |
UN Number | UN3399 |
Solubility Information | Soluble in water( 46g/100 mL (26°C). |
Formula Weight | 160.78 |
Percent Purity | 97% |
Physical Form | Liquid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'