missing translation for 'onlineSavingsMsg'
Learn More

Triethylenetetramine, tech. 60%, balance branched and cyclic triethylenetetramines

Product Code. 11473167
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11473167 2500 mL 2500mL
11463167 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11473167 Supplier Thermo Scientific Alfa Aesar Supplier No. A16128.0F
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 112-24-3 | C6H18N4 | 146.238 g/mol

Catalyzes the hydrolysis of N-terminal peptide bonds. Determination of the cation exchange capacity (CEC) of clay minerals is by using the complexes of copper(II) ion with triethylenetetramine and tetraethylenepentamine. Variable spin iron(iii) chelates with hexadentate ligands are derived from triethylenetetramine and various salicylaldehydes. Adsorption of Cu(II), Cd(II) and Pb(II) from aqueous single metal solutions by succinylated twice-mercerized sugarcane bagasse functionalized with triethylenetetramine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Catalyzes the hydrolysis of N-terminal peptide bonds. Determination of the cation exchange capacity (CEC) of clay minerals is by using the complexes of copper(II) ion with triethylenetetramine and tetraethylenepentamine. Variable spin iron(iii) chelates with hexadentate ligands are derived from triethylenetetramine and various salicylaldehydes. Adsorption of Cu(II), Cd(II) and Pb(II) from aqueous single metal solutions by succinylated twice-mercerized sugarcane bagasse functionalized with triethylenetetramine.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-24-3
Molecular Formula C6H18N4
Molecular Weight (g/mol) 146.238
MDL Number MFCD00008169
InChI Key VILCJCGEZXAXTO-UHFFFAOYSA-N
Synonym triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951
PubChem CID 5565
ChEBI CHEBI:39501
IUPAC Name N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine
SMILES C(CNCCNCCN)N

Specifications

Melting Point ∼-35°C
Density 0.982
Boiling Point ∼267°C
Flash Point 122°C (252°F)
Odor Amine-like
Linear Formula H2NCH2CH2(NHCH2CH2)2NH2
Refractive Index 1.498
Quantity 2500 mL
UN Number UN2259
Beilstein 605448
Sensitivity Hygroscopic
Merck Index 14,9663
Solubility Information Insoluble in water.
Formula Weight 146.24
Percent Purity 60%
Grade Technical
Assay balance branched and cyclic triethylenetetramines
Chemical Name or Material Triethylenetetramine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.