missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triethyleneglycol Monolauryl Ether-d25, TRC
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
Specifications
Specifications
| Chemical Name or Material | Triethyleneglycol Monolauryl Ether-d25 |
| CAS | 171922-68-2 |
| Molecular Formula | C18H13D25O4 |
| Quantity | 25 mg |
| Synonym | 3,6,9-Trioxaheneicosan-1-ol-d25, 2-[2-[2-(Dodecyloxy)ethoxy]ethanol-d25, C12EO3-d25, Dodecyl Triethylene Glycol-d25, Ethal 326, LEA-d25, Laureth-3-d25, Lauryl Alcohol Triglycol Ether-d25, Lauryl Triethoxylate-d25, Lauryl Triglycol Ether-d25, Nikkol BL 3SY-d25, Triethylene Glycol Dodecyl Ether-d25, Triethylene Glycol Lauryl Ether-d25, Triethylene Glycol Monododecyl Ether-d25, Tris(oxyethylene) Dodecyl Ether-d25, n-Dodecyl Triethylene Glycol Ether-d25 |
| InChI Formula | InChI=1S/C18H38O4/c1-2-3-4-5-6-7-8-9-10-11-13-20-15-17-22-18-16-21-14-12-19/h19H,2-18H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,13D2 |
| SMILES | [2H]C([2H])(C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])OCCOCCOCCO |
| Molecular Weight (g/mol) | 343.65 |
| Formula Weight | 343.4339 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction