missing translation for 'onlineSavingsMsg'
Learn More

Triethylene Glycol Ditosylate, TRC

Product Code. 30370570
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30370570 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30370570 Supplier Toronto Research Chemicals Supplier No. TRCE91774350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (Ethane-1,2-diylbis(oxy))bis(ethane-2,1-diyl) bis(4-Methylbenzenesulfonate)
CAS 19249-03-7
Molecular Formula C20H26O8S2
Quantity 50 mg
Synonym Triethyleneglycol bis(toluenesulfonate), (3,6-Dioxaoctane-1,8-diyl)bis(4-methyl-benzenesulfonate), 1,2-Ethanediylbis(oxy-2,1-ethanediyl) bis(4-methylbenzenesulfonate), 1,8-Bis(p-tolylsulfonyloxy)-3,6-dioxaoctane, 1,8-Bis(tosyloxy)-3,6-dioxaoctane, 2-[2-[2-[(4-Methylphenylsulfonyl)oxy]ethoxy]ethoxy]ethyl 4-methylbenzene-1-sulfonate, 2-[2-[2-[[(4-Methylphenyl)sulfonyl]oxy]ethoxy]ethoxy]ethyl 4-methylbenzenesulfonate, 3,6-Dioxaoctane-1,8-diol Ditosylate, NSC 244980, Triethylene Glycol Bis(4-toluenesulfonate), Triethylene Glycol Bis(p-toluenesulfonate), Triethylene Glycol Bis(p-tosylate), Triethylene Glycol Di-4-toluenesulfonate, Triethylene Glycol Di-p-tosylate, Triethylene Glycol Ditoluenesulfonate
InChI Formula InChI=1S/C20H26O8S2/c1-17-3-7-19(8-4-17)29(21,22)27-15-13-25-11-12-26-14-16-28-30(23,24)20-9-5-18(2)6-10-20/h3-10H,11-16H2,1-2H3
SMILES CC1=CC=C(S(=O)(OCCOCCOCCOS(=O)(C2=CC=C(C=C2)C)=O)=O)C=C1
IUPAC Name 2-[2-[2-(p-tolylsulfonyloxy)ethoxy]ethoxy]ethyl 4-methylbenzenesulfonate
Molecular Weight (g/mol) 458.54
Formula Weight 458.1069
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.