missing translation for 'onlineSavingsMsg'
Learn More

Triethylene glycol dimethyl ether, 99%

Product Code. 11386796
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11386796 100 g 100g
11396796 500 g 500g
11306806 2500 g 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11386796 Supplier Thermo Scientific Alfa Aesar Supplier No. L04128.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-49-2 | C8H18O4 | 178.228 g/mol

Triethylene glycol dimethyl ether is used as a solvent for gases and coupling immiscible liquid. It is also used to prepare perfluoro-triethyleneglycoldimethylether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethylene glycol dimethyl ether is used as a solvent for gases and coupling immiscible liquid. It is also used to prepare perfluoro-triethyleneglycoldimethylether.

Solubility
Miscible with water, chloroform and methanol

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-49-2
Molecular Formula C8H18O4
Molecular Weight (g/mol) 178.228
MDL Number MFCD00008504
InChI Key YFNKIDBQEZZDLK-UHFFFAOYSA-N
Synonym triglyme, triethylene glycol dimethyl ether, 2,5,8,11-tetraoxadodecane, 1,2-bis 2-methoxyethoxy ethane, glyme 4, ansul ether 161, glyme-3, tegdime, tegdme, unii-32yxg88kk0
PubChem CID 8189
ChEBI CHEBI:44842
IUPAC Name 1-methoxy-2-[2-(2-methoxyethoxy)ethoxy]ethane
SMILES COCCOCCOCCOC

Specifications

Melting Point -45°C
Density 0.986
Boiling Point 215°C to 216°C
Flash Point 113°C (235°F)
Linear Formula CH3O(CH2CH2O)3CH3
Refractive Index 1.423
Quantity 100 g
Beilstein 1700630
Sensitivity Hygroscopic
Merck Index 14,9692
Solubility Information Miscible with water,chloroform and methanol
Formula Weight 178.23
Percent Purity 99%
Chemical Name or Material Triethylene glycol dimethyl ether
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Triethylene glycol dimethyl ether
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H319-Causes serious eye irritation.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P233-Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Triethylene glycol dimethyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.