missing translation for 'onlineSavingsMsg'
Learn More

Triethylene glycol dimethyl ether, 99%, stabilized

Product Code. 10563504 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10563504 2.5 L Glass bottle 2.5 L
10657492 250 mL Glass bottle 250mL
10295280 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10563504 Supplier Thermo Scientific Acros Supplier No. 139600025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-49-2 | C8H18O4 | 178.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-49-2
Molecular Formula C8H18O4
Molecular Weight (g/mol) 178.23
MDL Number MFCD00008504
InChI Key YFNKIDBQEZZDLK-UHFFFAOYSA-N
Synonym triglyme, triethylene glycol dimethyl ether, 2,5,8,11-tetraoxadodecane, 1,2-bis 2-methoxyethoxy ethane, glyme 4, ansul ether 161, glyme-3, tegdime, tegdme, unii-32yxg88kk0
PubChem CID 8189
ChEBI CHEBI:44842
IUPAC Name 1-methoxy-2-[2-(2-methoxyethoxy)ethoxy]ethane
SMILES COCCOCCOCCOC

Specifications

Acidity 0.05mg KOH/g max.
Melting Point -45°C
Density 0.9860g/mL
Boiling Point 216°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OCH2CH2OCH2CH2OCH2CH2OCH3
Refractive Index 1.4220 to 1.424
Quantity 2.5 L
Merck Index 15, 9861
Specific Gravity 0.986
Solubility Information Solubility in water: soluble. Other solubilities: miscible with hydrocarbon solvents
Stabilizer 0.01% BHT
Viscosity 2.5 mPa.s (20°C)
Formula Weight 178.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Triethylene glycol dimethyl ether, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Triethylene glycol dimethyl ether, stabilized
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H319-Causes serious eye irritation.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P233-Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.