missing translation for 'onlineSavingsMsg'
Learn More

Triethylene Glycol Dimethyl Ether, 99%, Reagent Grade, Honeywell™

Product Code. 15678180 Shop All Solstice Research Chemicals Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
250 g
1 L
Packaging:
Glass bottle
missing translation for 'unitSize':
1 L
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging missing translation for 'unitSize'
15678180 250 g Glass bottle 250g
15658180 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 15678180 missing translation for 'mfr' Honeywell Research Chemicals missing translation for 'supplierNo' T59803250G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-49-2
Molecular Formula C8H18O4
Molecular Weight (g/mol) 178.228
MDL Number MFCD00008504
InChI Key YFNKIDBQEZZDLK-UHFFFAOYSA-N
Synonym triglyme, triethylene glycol dimethyl ether, 2,5,8,11-tetraoxadodecane, 1,2-bis 2-methoxyethoxy ethane, glyme 4, ansul ether 161, glyme-3, tegdime, tegdme, unii-32yxg88kk0
PubChem CID 8189
ChEBI CHEBI:44842
IUPAC Name 1-methoxy-2-[2-(2-methoxyethoxy)ethoxy]ethane
SMILES COCCOCCOCCOC

Specifications

Melting Point -45°C
Density 0.986g/mL at 25°C
Boiling Point 216°C
Flash Point 113°C (235.4°F)
Ignition Point 375°F
Packaging Glass bottle
Linear Formula CH3O(CH2CH2O)3CH3
Quantity 250 g
UN Number NONH for all modes of transport
Beilstein 1700630
Vapor Density >4.7 (vs air)
Formula Weight 178.23g/mol
Percent Purity 0.99
Chemical Name or Material Triethylene glycol dimethyl ether
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.