missing translation for 'onlineSavingsMsg'
Learn More

Triethylene Glycol Dimethacrylate (Stabilized with MEHQ), TRC

Product Code. 30450247
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30450247 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30450247 Supplier Toronto Research Chemicals Supplier No. TRCT7764705G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 109-16-0
Molecular Formula C14H22O6
Quantity 5 g
Synonym 2-Methyl-2-propenoic Acid 1,1'-[1,2-ethanediylbis(oxy-2,1-ethanediyl)] Ester, Methacrylic Acid Ethylenebis(oxyethylene) Ester, 1,2-Bis[2-(methacryloyloxy)ethoxy]ethane, ATM 2, Acryester 3ED, Bisomer TEGDMA, Blemmer PDE 150, EM 328, Esschem 943X7469, Ethylenebis(oxyethylene) methacrylate, NSC 84260, Neomer PM 201, SR 205, Sartomer SR 205, TEDMA, TEGDMA, TGM 3, TGM 3S
InChI Formula InChI=1S/C14H22O6/c1-11(2)13(15)19-9-7-17-5-6-18-8-10-20-14(16)12(3)4/h1,3,5-10H2,2,4H3
SMILES CC(=C)C(=O)OCCOCCOCCOC(=O)C(=C)C
IUPAC Name 2-[2-[2-(2-methylprop-2-enoyloxy)ethoxy]ethoxy]ethyl 2-methylprop-2-enoate
Molecular Weight (g/mol) 286.32
Formula Weight 286.1416
Chemical Name or Material Triethylene Glycol Dimethacrylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.